Learn More
OGT Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B2A6, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | OGT |
---|---|
Clone | 2B2A6 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849055
|
Proteintech
66823-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16839055
|
Proteintech
66823-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
O-linked N-acetylglucosamine (O-GlcNAc), is a member of the O-GlcNAc transferase family. O-GlcNAc is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine/threonine hydroxyl groups. It plays an important role in nutrient sensing, gene expression, and protein degradation.Specifications
OGT | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FLJ23071, HRNT1, O GLCNAC, OGT | |
OGT | |
IgG2a | |
Protein A | |
Antibody |
2B2A6 | |
Western Blot | |
Unconjugated | |
Mouse | |
Pig, Human, Rat, Mouse | |
O15294, P56558, Q27HV0 | |
108155, 26295, 664652, 8473 | |
OGT Fusion Protein Ag28402 | |
Primary | |
-20°C | |
OGT |