Learn More
Masterflex™ Nylon Male Pipe Adapter
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30621-63
This item is not returnable.
View return policy
Description
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Barb x NPT(M) Connection | |
Nylon | |
3/8 in. | |
3/8 in. | |
3/8 in. NPT(M) | |
Male Pipe Adapter |
Not Autoclavable | |
3/8 in. | |
3/8 in. NPT(M) | |
Ethylene Oxide | |
Translucent |