Learn More
Masterflex™ Nylon Female Luer Fitting
Use these low-profile hose barbs with semi-rigid tubing such as nylon, polyethylene and polypropylene. Nylon Female Luer Fittings feature a smooth bore hose barb to reduce flow turbulence and risk of line contamination.
136.00 NOK - 677.00 NOK
Specifications
Material | Nylon |
---|---|
Packaging Quantity | 25 Pack |
Product Type | Female Luer Fitting |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
15101425
|
Masterflex™
45502-03 |
Female Luer x 3/32 in. Wide Bore Hose Barb | N/A | ||||||
15164027
|
Masterflex™
45505-80 |
Female Luer x 1/8-27 NPT Elbow |
677.00 NOK
Pack of 25 |
Estimated Shipment: 02-02-2026
Log in to see stock. |
|||||
13525770
|
Masterflex™
45502-02 |
Female Luer x 3/32 in. Hose Barb |
200.00 NOK
Pack of 25 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
|||||
15351161
|
Masterflex™
45502-05 |
Female Luer x 1/8 in. Wide Bore Hose Barb | N/A | ||||||
Description
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
Specifications
Nylon | |
25 Pack | |
Female Luer Fitting |