Learn More
Masterflex™ Nylon Barbed NPT Female Pipe Adapters
Designed for quick and easy assembly without tools
Brand: Masterflex™ 31500-40
This item is not returnable.
View return policy
Description
- Combine these simple and economical fittings with flexible tubing.
- For use in low-pressure applications (generally less than 30psi).
Specifications
Barb x NPT(F) Connection | |
Ethylene Oxide | |
Barbed NPT female pipe adapters, Nylon, 1//8 in. NPT x 1/8 in. ID, 1/16 in., 1-1/32 in., 5/8 in. |
Nylon | |
16 mm |