Learn More
Masterflex™ Nylon Barbed Elbow Pipe Adapter
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30623-05
This item is not returnable.
View return policy
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Barb x NPT(M) Connection | |
3/8 in. | |
3/8 in. NPT(M) | |
3/8 in. NPT(M) |
Not Autoclavable | |
3/8 in. | |
Ethylene Oxide | |
Barbed Fitting |
Product Content Correction
Product Title
Masterflex™ Nylon Barbed Elbow Pipe Adapter > Nylon Barbed Elbow Pipe Adapter