Learn More
Masterflex™ Nylon Barbed Elbow Pipe Adapter
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30623-05
Product Code. 11780149
  • 287.00 NOK / Pack of 10
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
Barb x NPT(M) Connection
3/8 in.
3/8 in. NPT(M)
3/8 in. NPT(M)
Not Autoclavable
3/8 in.
Ethylene Oxide
Barbed Fitting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Nylon Barbed Elbow Pipe Adapter > Nylon Barbed Elbow Pipe Adapter