Learn More
Macherey-Nagel™ NucleoSpin™ DNA Stool, Mini kit for DNA from stool
Combines enzymatic lysis with mechanical disruption of cell walls for efficient isolation of high-quality genomic DNA from any stool sample, fresh or frozen. Complete kit includes NucleoSpin™ Bead Tubes.
Brand: Macherey-Nagel™ 740472.50
3497.94 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Isolation of genomic DNA from bacterial and epithelial cells from stool samples
- Efficient procedure combining enzymatic lysis and bead disruption
- Isolation of high-quality genomic DNA (microbial and host) from fresh and frozen samples
- Suitable for any stool sample – carnivores, omnivores, and herbivore
- Bead Tubes included for efficient lysis – compatible with most common disruption devices
- NucleoSpin™ Inhibitor Removal Columns ensure highest purity of the isolated DNA
- Downstream applications: PCR, real-time PCR, Southern blotting, microarray technology
Specifications
Room temperature (18 - 25 °C) | |
Silica MembraneTechnology and Mechanical (Bead Tube) Disruption | |
Binding capacity: 50 μg | |
Disruption device (e.g. Vortex-Genie 2 with MN Bead Tube Holder), Centrifuge, Vortex mixer | |
DNA Stool Columns, Bead Tubes Type A, Inhibitor Removal Columns, Collection Tubes (2 mL), Collection Tubes (2 mL, lid), Buffers | |
A260/A280: 1.7 - 1.9 | |
Fragment size: 200 bp - approx. 50 kbp | |
Varies by sample type, quality and water content |
Mini Spin Column | |
Stool Samples (Herbivores, Omnivores, or Carnivores, Fresh or Frozen) | |
Elution volume: 30 - 100 μL | |
Isolation of genomic DNA from stool samples | |
50 | |
Approx. 200 mg (180 - 220 mg) | |
60 min/10 prep |
Safety and Handling
- NucleoSpin DNA Stool
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash with water thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P330-Rinse mouth.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.