Learn More
Macherey-Nagel™ Nucleodur™ C18 Varioprep VP HPLC Columns
Column Format: Preparative Column; Carbon Load: 18%; Pore Size: 110 Å; Column Phase: Reversed Phase
9835.00 NOK - 12235.00 NOK
Specifications
Column Format Preparative Column
Diameter (Metric) 10 mm
For Use With Sophisticated Analytical and Preparative Separations of Basic, Neutral and Acidic Pharmaceuticals, Derivatized Amino Acids, Pesticides, Fat-Soluble Vitamins, Aldehydes, Ketones and Phenolic Compounds
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
12616009
View Documents
Macherey-Nagel™
762554.100
125 mm
9835.00 NOK
Each
Only null left
Estimated Shipment: 18-07-2025
Add to basket
12626009
View Documents
Macherey-Nagel™
762556.100
250 mm
12235.00 NOK
Each
Only null left
Estimated Shipment: 18-07-2025
Specifications
Preparative Column
Sophisticated Analytical and Preparative Separations of Basic, Neutral and Acidic Pharmaceuticals, Derivatized Amino Acids, Pesticides, Fat-Soluble Vitamins, Aldehydes, Ketones and Phenolic Compounds
5 μm
110 Å
60°C
C18 HT
18%
Yes
Preparative Column
10 mm
5800 psi (400 bar)
1 to 11
340 m²/g
L1
Silica
Reversed Phase
Reversed Phase