Learn More
Invitrogen™ Novex™ Semi-Dry Blotter
Quickly and efficiently transfers proteins and nucleic acids from mini and midi gels to membranes
Brand: Invitrogen™ SD1000
Requires : External power supply for operation (up to 800mA, depending on the number and size of blots).
Description
Simultaneous transfer can be achieved from up to 2 midi gels, 4 mini gels, or 2 E-PAGE™ gels. The Novex™ Semi-Dry Blotter is designed for fast set-up and ease of use. The platinum-coated titanium anode plate and stainless steel cathode plate provide fast and efficient electroblotting using less transfer buffer than wet (tank) blotting methods. The Novex™ Semi-Dry Blotter is designed for convenience and efficiency.
- Economical – Horizontal configuration requires less than 200mL of buffer to transfer
- High-capacity – A large, 20 x 20cm transfer surface allows up to two midi gels or four mini gels to be transferred at one time
- Flexible – Field strength across the blot is maximized and evenly distributed, for efficient and complete transfers regardless of the number and size of gels blotted
- Durable – Stainless steel and platinum-coated titanium electrodes allow for corrosion-free operation
- Fast – Blotting is typically completed in 30 to 60 min
- Compatible with Novex™ midi and mini gels and NuPAGE™ gels
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. | |
Western Blot | |
NuPAGE™ Gel, Novex™ Midi Gel, Novex™ Mini Gel | |
Nitrocellulose, PVDF | |
Novex | |
Transfer Device | |
Horizontal |
2 E-PAGE Gels, 2 Midi-Gels,4 Mini-Gels | |
Novex™ Semi-Dry Blotter | |
Mini 8 cm x 8 cm, Midi 8 cm x 13 cm | |
Semi-Dry | |
2 Midi or 4 Mini Gels | |
Each | |
Room Temperature |