Learn More
Nickel Copper foil, alloy 400, 1.98mm (0.078 in.) thick, Thermo Scientific™
1816.89 NOK - 8190.00 NOK
Specifications
Chemical Name or Material | Nickel Copper foil, Alloy 400 |
---|---|
Name Note | 1.98mm (0.078 in.) thick |
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cu; 67:33 wt% |
MDL Number | MFCD00801098 |
Form | Foil |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15406805
|
Thermo Scientific Alfa Aesar
045170.GT |
150 x 150 mm | N/A | ||||||
15416805
|
Thermo Scientific Alfa Aesar
045170.HB |
300 x 300 mm |
8190.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nickel Copper foil, Alloy 400 | |
Ni:Cu; 67:33 wt% | |
Foil |
1.98mm (0.078 in.) thick | |
MFCD00801098 |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures