missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26003-1-AP-150UL
This item is not returnable.
View return policy
Description
26003-1-AP specifically recognizes the 20 kDa protein.
NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene are a cause of mitochondrial complex I deficiency.Specifications
NDUFAF4 | |
Polyclonal | |
Unconjugated | |
NDUFAF4 | |
C6orf66, HRPAP20, HSPC125, My013, NDUFAF4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29078 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9P032 | |
NDUFAF4 | |
NDUFAF4 Fusion Protein Ag23365 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction