missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine trihydrofluoride, 90%

Product Code. p-3559331 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Plastic bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 131600-43-6 | C8H22F3N | 189.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131600-43-6
Molecular Formula C8H22F3N
Molecular Weight (g/mol) 189.27
MDL Number MFCD00144880
InChI Key AJRRXKJZYYBJPY-UHFFFAOYSA-N
Synonym diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-amine trihydrofluoride, n,n-diisopropylethylamine trihydrofluoride, acmc-1bx1e, ksc490i4d, n,n,-diisopropylethylamine trihydrofluoride, n-ethyl-n-isopropylpropan-2-aminetrihydrofluoride, n-ethyl-n-isopropyl-propan-2-amine trihydrofluoride, n-ethyl-n-propan-2-yl-2-propanamine trihydrofluoride, n-ethyl-n-propan-2-yl-propan-2-amine trihydrofluoride
PubChem CID 45108321
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine;trihydrofluoride
SMILES F.F.F.CCN(C(C)C)C(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.9650g/mL
Boiling Point 93°C to 95°C (4.0 mmHg)
Flash Point 40.5°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Plastic bottle
Quantity 1 g
Specific Gravity 0.965
Solubility Information Solubility in water: soluble
Formula Weight 189.26
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material N, N-Diisopropylethylamine trihydrofluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Diisopropylethylamine trihydrofluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.