Learn More
Wheaton™ Motor
Brand:  Wheaton™ I052713
16608.35 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12893016
  • 19125.00 NOK / Each
Only null left
Estimated Shipment: 09-01-2026
Specifications
28 rpm
90 VDC
Shaft: 7.9 mm