missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum solution for ICP 1g/L NH3 matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
667.00 NOK - 1370.00 NOK
Specifications
Analyte or Component Names | Mo |
---|---|
Concentration | 1±0.002 g/L |
Type | Mass Spectrometry Standard |
Element | Mo |
Solvent or Matrix | NH3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12916204
![]() |
Fisher Chemical
S 134 100 |
100 mL | Bottle |
667.00 NOK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12926204
![]() |
Fisher Chemical
S 134 250 |
250 mL | Bottle |
1075.00 NOK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12936204
![]() |
Fisher Chemical
S 134 500 |
500 mL | Bottle |
1370.00 NOK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Mo | |
Mass Spectrometry Standard | |
NH3 |
1±0.002 g/L | |
Mo |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 months (12 months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Molybdenum (Mo) 1000mg/l in NH3 4%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title