Learn More
Cytiva™ MiniTrap™ Column
Shop All Cytiva Products
Click to view available options
Quantity:
1 Ea.
Unit Size:
Pack of 50
Description
- For rapid and efficient separation and purification of samples
- Single use, pre-packed, disposable format allows for easy and convenient sample processing
- Ideal for use in R and D, process development and QC laboratories
Specifications
Specifications
Description | MiniTrap chromatography column |
Product Type | Column |
For Use With (Application) | Small-scale purification and separation |
Format | Chromatography Cartridge |
Disposable | Single-use |
Quantity | 1 Ea. |
Safety and Handling
Signal Word
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 5-chloro-2-methyl-2H-isothiazol-3-one