Learn More
Grant Instruments™ Microtube Insert
Grant Instruments Microtube insert used with MPS-1 High-speed shaker/vortex mixer.
Brand:  Grant Instruments™ P-2-24
690.23 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15379625
  • 713.00 NOK / Each
Only null left
Estimated Shipment: 07-10-2025
Description
  • Made from Polyethylene Foam EVA PE140
  • Color: Blue
  • For use with MPS-1 High-speed shaker/vortex mixer
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    24 x 1.5/2.0 mL Tubes
    Polyethylene Foam EVA PE140 Base
    MPS-1 High-Speed Shaker/Vortex Mixer
    1.5/2 mL