Learn More
Ohaus™ Microtube Holder Kit with Retainer
OHAUS™ Microtube Holder Kit with Retainer for effortlessly mixing microtubes and microplates.
Brand: Ohaus™ 30400200
123.99 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Microtube Holder Kit | |
Foam | |
Microtube Kit with Retainer |