Learn More
Ansell™ Microgard™ FR Coveralls with Hood
Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Coverall with 2-piece hood, stitched seams, elasticated hood, wrists, waist and ankles
- 2-way front zipper with storm flap
- Flame retardant treated Sontara/wood pulp/polyester fabric
- 3-thread overlocked seams
Specifications
Specifications
Product Type | Hooded Flame-resistant Coveralls |
Size | X-Large |
Material | Multi-layer Barrier Laminate Fabric |
Color | Blue |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 2-panel |
Closure Type | 2-way Front Zipper with Storm Flap |