Learn More
Ansell Microgard™ 2000 Standard White Coveralls with Hood
Allows perspiration to escape from the suit yet withstands saturation of liquid chemicals. Ansell Microgard™ 2000 Standard White Coveralls with Hood filter 100% of particulates down to 0.01 microns in size.
Brand: Ansell WH20B-00111-04
3401.33 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Coverall with 3-piece hood
- White bound seams
- Elasticated hood, waist, wrists and ankles
- Includes fingerloops
- 2-way front zipper with re-sealable storm flap.
- Liquid penetration resistance and barrier to fine particulates (>0.01 microns)
- High quality two-way stretch microporous film provides an effective liquid and particle barrier
- Moisture vapor permeable to help reduce the risk of heat stress
- Silicone free - Critical in paint spraying applications
- Low linting - Reduces the risk of fibre contamination in some critical areas
- Optimized body fit provides wearer comfort and safety
- Anti-static treatment
Specifications
General Purpose Coveralls | |
Unisex | |
White | |
Elastic | |
Thumb Loop | |
3-panel, Elastic | |
Bound | |
Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781 | |
None |
Large | |
Microporous Polyethylene Film | |
Single-use | |
Bound | |
Hooded | |
Front Zipper with Storm Flap | |
Non-sterile | |
WH20-B-00-111 | |
Includes Hood, Fingerloops |