Learn More
Ansell Microgard™ 1500 Plus White Coveralls with Hood
Provides a good barrier against particulates such as asbestos, brick dust and cement dust. Ansell Microgard™ 1500 Plus White Coveralls with Hood filter 99.9% of particles >3μ.
Brand: Ansell WH15S-00111-06
1573.33 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Coverall with 3-piece hood, stitched seams, elasticated hood, waist, wrists and ankles
- 2-way front zipper with re-sealable stormflap
- Anti-static treated SMS fabric
- Breathable to help reduce the risk of heat stress
- Silicone-free which is critical in spray painting applications
- Optimized body fit for wearer comfort and safety
Specifications
General Purpose Coveralls | |
Unisex | |
White | |
Elastic | |
Elastic | |
3-panel, Elastic | |
Stitched | |
EN ISO 13982-1, EN13034, EN1073-2, EN1149-5 | |
None |
2X-Large | |
Anti-static SMS | |
Single-use | |
Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Non-sterile | |
WH15-S-00-111 | |
Includes Hood |