Learn More
Ansell™ Microchem™ 4000 Apollo Encapsulated Coveralls
Trusted by fire and rescue crews around the world. MICROCHEM™ 4000 APOLLO is a fully encapsulated liquid tight chemical suit designed for use in conjunction with self contained breathing apparatus (SCBA)
11545.94 NOK
Specifications
Certifications/Compliance | Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5 |
---|---|
Color | Green |
Hood Type | Fully Encapsulated |
Material | Multi-layer Barrier Laminate |
Ankle Style | Elastic, Attached socks, Boot overflaps |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15546454
|
Ansell™
GR40T-00126-03 |
Medium |
11545.94 NOK
Pack of 4 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
15556454
|
Ansell™
GR40T-00126-04 |
Large |
11545.94 NOK
Pack of 4 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
15566454
|
Ansell™
GR40T-00126-05 |
X-Large |
11545.94 NOK
Pack of 4 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
15576454
|
Ansell™
GR40T-00126-06 |
2X-Large |
11545.94 NOK
Pack of 4 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
Description
- Full Encapsulated coverall
- Rear double-zip entry
- Expanded back for internal wearing of self-contained breathing apparatus (SCBA)
- Rear positioned exhalation valves
- Internal braces
- Escape “rip-cord” for rapid doffing in emergency situations
- Attached socks with boot over flap
- Attached Ansell Barrier™ gloves with sleeve over flap
Specifications
Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5 | |
Fully Encapsulated | |
Elastic, Attached socks, Boot overflaps | |
Attached gloves | |
None | |
Welded, Taped |
Green | |
Multi-layer Barrier Laminate | |
Rear-entry double zipper | |
Hooded | |
Encapsulated coveralls | |
Includes Attached Ansell Barrier™ Gloves, integrated socks with boot overflap, fully encapsulated hood |