Learn More
Ansell™ Microchem™ 3000 Coveralls with Hood and Integrated Socks
MICROCHEM™ 3000 Coveralls with Hood and Integrated Socks is one of the lightest and most comfortable chemical protective garments on the market today. This durable multi-layer fabric provides an extremely effective barrier against both inorganic chemicals and biological hazards.
Brand: Ansell™ YE30W-00122-04
7264.51 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Coverall with 2-piece hood
- Integrated socks with boot overflap
- Double zip closure
- Double cuffs
- Elasticated hood, waist, double cuffs
- Excellent liquid penetration resistance and barrier to fine particulates (>0.01 microns)
- Moisture vapour permeable (“breathable”) to help reduce the risk of heat stress
- Silicone free for paint spraying applications
- Low linting to reduces the risk of fibre contamination in some critical areas
- Optimised body fit for wearer comfort and safety
- Anti-static according to EN 1149-5
Specifications
Protective Coveralls | |
Multi-layer Barrier Laminate Fabric | |
Elastic | |
Double, Elastic | |
2-panel, Elastic | |
Welded | |
Hood, integrated socks with boot overflap | |
Includes Hood, Integrated socks with boot overflap |
Large | |
Yellow | |
Elastic, Attached Socks, Boot Overflaps | |
Hooded | |
Double Zipper | |
Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5 | |
None |