Learn More
Ansell™ Microchem™ 3000 Aprons with Sleeves
Constructed of a multi-layer barrier laminate. Ansell Microchem™ 3000 Aprons with Sleeves with welded seams provide liquid tight connections.
11785.00 NOK - 13250.00 NOK
Specifications
Material Multi-layer Barrier Laminate Fabric
Color Yellow
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15596594
View Documents
Ansell™
YE30W-00214-02
Small
13250.00 NOK
Pack of 45
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 25-11-2025
Add to basket
15506604
View Documents
Ansell™
YE30W-00214-03
Medium
13250.00 NOK
Pack of 45
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 28-08-2025
Add to basket
15516604
View Documents
Ansell™
YE30W-00214-04
Large
13250.00 NOK
Pack of 45
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 11-09-2025
Add to basket
15526604
View Documents
Ansell™
YE30W-00214-05
X-Large
13250.00 NOK
Pack of 45
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 25-11-2025
Add to basket
15536604
View Documents
Ansell™
YE30W-00214-06
2X-Large
11785.00 NOK
Pack of 40
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 25-11-2025
Add to basket
17111115
View Documents
Ansell™
YE30W-00214-07
3X-Large
11785.00 NOK
Pack of 40
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 16-09-2025
Add to basket
Description
Description
  • Apron with Sleeves
  • Rear velcro fastening
  • Elasticated wrists
  • Welded seams
  • Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible bright yellow for improved worker safety
  • Lightweight yet durable
  • Anti-static according to EN 1149-5
Specifications
Specifications
Multi-layer Barrier Laminate Fabric