Learn More
MFN2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28341-1-AP-150UL
Description
The MFN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MFN2. This antibody recognizes human, mouse, rat antigen. The MFN2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
MFN2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
9927 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
467 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |