Learn More
Methoxydimethylphenylsilane, 97%, Thermo Scientific™
Product Code. 11406310
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17881-88-8
Molecular Formula C9H14OSi
Molecular Weight (g/mol) 166.295
MDL Number MFCD09266142
InChI Key REQXNMOSXYEQLM-UHFFFAOYSA-N
Synonym methoxydimethylphenylsilane, methoxydimethyl phenyl silane, silane, methoxydimethylphenyl, dimethylmethoxyphenylsilane, dimethylphenylmethoxysilane, benzene, methoxydimethylsilyl, acmc-1boft, phenyldimethylmethoxysilane, amtsi001
PubChem CID 140300
IUPAC Name methoxy-dimethyl-phenylsilane
SMILES CO[Si](C)(C)C1=CC=CC=C1
Specifications
Boiling Point 82°C (28mmHg)
Quantity 5 g
Formula Weight 166.3
Percent Purity 97%
Sensitivity Moisture sensitive
Chemical Name or Material Methoxydimethylphenylsilane