Learn More
3M™ Metaliks™ Sport Safety Glasses
Click to view available options
Lens Tint:
Amber
Blue, Mirror
Unit Size:
Pack of 20
Description
- Semi-rimless metal frame and optimal field of vision
- Soft-touch temple tips provide additional comfort
- Soft saddle nose pad for enhanced comfort
- Optical class 1 for maximum clarity
- Anti-Scratch and Anti-Fog coating for increased durability and improved vision
- Soft-touch temple tips provide additional comfort
- Blue Mirror coating for increased durability and improved vision
Specifications
Specifications
Certifications/Compliance | EN 166: 2001 |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Amber |
Product Type | Safety glasses |
For Use With (Application) | Construction, forestry |
Frame Color | Silver |
Lens Material | Polycarbonate |
Frame Material | Metal |