missing translation for 'onlineSavingsMsg'
Learn More

Melanin, MP Biomedicals™

Product Code. 11417632 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 8049-97-6
Molecular Formula C18H10N2O4
Molecular Weight (g/mol) 318.288
InChI Key XUMBMVFBXHLACL-UHFFFAOYSA-N
Synonym melanin, phaeomelanins, d0wq4g, melanin from sepia officinalis, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1; 2,5 .0; 13,17 .0; 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone, 6,14-dimethyl-4,12-diazapentacyclo 8.6.1.1^ 2,5 .0^ 13,17 .0^ 9,18 octadeca-1 17 ,2,5,9 18 ,10,13-hexaene-7,8,15,16-tetrone
PubChem CID 6325610
SMILES CC1=C2C3=C(C4=CNC5=C(C(=O)C(=O)C(=C45)C3=CN2)C)C(=O)C1=O

Specifications

Color Brown to Black
Quantity 1 g
Formula Weight 1780
Physical Form Granular Powder
Chemical Name or Material Melanin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.