Learn More
Marble Chips, Pure, Fisher Chemical™
Product Code. 10284530 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
3 kg
Marble Chips, Pure
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O
Specifications
Melting Point 825°C
Color White
pH 8 to 9
Physical Form Solid
Quantity 1 kg
Formula Weight 100.09g/mol
Packaging Plastic powder jar
Product Identifier
  • Calcium carbonate