Learn More
Hamilton™ Male Luer Lock M6 Threads
Hamilton offers a diverse line of inert fittings fittings and connectors made from CTFE. The fittings are designed for use with industry standard luer and luer lock products like our syringes as well as threaded 1/4-28 and M6 flat bottom ports.
Brand: Hamilton™ 32876
678.53 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Specification Sheet
- Connection 1 – Male Luer Lock
- Connection 2 – Male M6
- Sales Quantity – 1 each
- I.D. – 1.5 mm
Specifications
Luer Lock Connector | |
Male Luer Lock M6 Threads |