missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium powder, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
25 kg
Unit Size:
100g
25kg
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Specifications
Specifications
Appearance | Complying |
Quantity | 100 g |
Percent Purity | ≥99% |
Packaging | Plastic Bottle |
Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction