Learn More
Micronova™ M-Zone™ Polyester Snap Mop
BIOPHARMAGREENER_CHOICE
Polyester mop with waffle weave for increased absorbency. Micronova™ M-Zone™ Polyester Snap Mop is compatible with strong disinfectants.
Brand:  Micronova™ MZSM11-14IR
6052.51 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15551755
  • 6795.00 NOK / Pack of 24
Only null left
Estimated Shipment: 03-11-2025
Description
  • Irradiated
  • Autoclavable
  • For use with QDSL and QPSL series mop adapters
  • Designed for clean and controlled environment
  • 100% Polyester with waffle weave for increased absorbency
  • Compatible with strong disinfectants
  • Autoclavable
Certifications
ISO 9001:2008
Compliance
AMI Guidelines for Sterilization
Specifications
Specifications
Polyester