Learn More
Buerkle™ LPDE Safety Wash Bottles with Leakproof Venting Valve
The venting valve ensures that the excess pressure can escape and dripping is prevented. With leakproof venting valve, contents cannot leak when bottle has fallen over as with wash bottles with ball valve or venting hole.
Brand: Buerkle™ 0310-2052
This item is not returnable.
View return policy
Description
- Safe filling due to wide-necked opening 39 mm neck diameter
- Permanent print according to GHS with substance name in German, English, French and Spanish, GHS pictograms and CAS number
- LDPE yellow transparent with light protection
- Contents 500 ml
Specifications
500 mL | |
LDPE | |
Yes | |
Methanol |
Transparent Yellow | |
Red | |
Safety Wash Bottle | |
Screw Cap |