Learn More
Lenz Laborglasintrumente Long Ground Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
619.00 NOK - 2750.00 NOK
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
10355021
|
Lenz Laborglasintrumente
01100714 |
NS 14/35 |
619.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10101951
|
Lenz Laborglasintrumente
01100734 |
NS 34/45 |
2085.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10204291
|
Lenz Laborglasintrumente
01100710 |
NS 10/30 |
884.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10234431
|
Lenz Laborglasintrumente
01100745 |
NS 45/50 |
2750.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10264331
|
Lenz Laborglasintrumente
01100719 |
NS 19/38 |
667.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10569781
|
Lenz Laborglasintrumente
01100729 |
NS 29/42 |
912.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10682272
|
Lenz Laborglasintrumente
01100705 |
NS 5/20 |
1750.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10769021
|
Lenz Laborglasintrumente
01100724 |
NS 24/40 |
888.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10788641
|
Lenz Laborglasintrumente
01100707 |
NS 7/25 |
1750.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10798641
|
Lenz Laborglasintrumente
01100712 |
NS 12/32 |
715.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
Description
- Conical ground cones
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |