Learn More
JULABO™ Lift-up Makrolon™ Bath Cover for SW22/23
Prevent liquid losses due to evaporation, protects samples from contamination to 80°C, for SW22/23.
Brand: JULABO™ 8970288
3193.77 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
This accessory is suitable for JULABO™ SW22, SW23Specifications
SW22, SW23, TW20 shaking water bath | |
Bath Cover | |
Prevent liquid losses due to evaporation, protects samples from contamination to 80°C | |
Makrolon |