Learn More
BINDER™ Left Access Port (30 mm)
For Use With: FDL 115
Brand: BINDER™ 8012-1360
6059.97 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 30 mm, left | |
Steel | |
Silicone plug |
BINDER Chambers FDL 115 | |
Access Port | |
30 mm |