Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class E2
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand: Fisherbrand™ FB67831
3440.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Class E2 stainless steel calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
E2 | |
1 to 500 mg | |
1 mg to 500 mg |