missing translation for 'onlineSavingsMsg'
Learn More

Lead titanium oxide, polymeric precursor, oxide >30 wt%, Thermo Scientific™

Product Code. 11324737
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in electronic devices

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-00-3
Molecular Formula O3PbTi
Molecular Weight (g/mol) 303.064
MDL Number MFCD00011159
InChI Key NKZSPGSOXYXWQA-UHFFFAOYSA-N
Synonym lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder,
PubChem CID 16211560
IUPAC Name dioxido(oxo)titanium;lead(2+)
SMILES [O-][Ti](=O)[O-].[Pb+2]

Specifications

Color Brown
Physical Form Viscous Liquid
Quantity 250 g
UN Number UN1993
Flash Point 12°C (54°F)
Solubility Information Soluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide >30 wt%
Sensitivity Moisture sensitive
Chemical Name or Material Lead titanium oxide, Polymeric precursor
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.