Learn More
Brand™ LDPE Wash Bottles without Valve
Shop All Brand Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Neutral
Closure Color:
Blue
Green
Orange
Red
White
Yellow
Description
- Manufactured from flexible LDPE
- Wide-mouth.
- High-quality wash bottles with integrated wash stream and sealing cone closure.
- Large opening for convenient filling.
- Label with designation of contents and general safety statements in English (with CAS numbers and NFPA codes) and German (with CAS numbers).
Specifications
Specifications
Capacity (Metric) | 500 mL |
Material | LDPE |
Closure Color | Green |
Mouth | Wide |
Product Type | Wash Bottle |
Chemical Label | Methanol |
Closure Type | Sealing Cone |