Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Warning; Crushing of hands
Sign compliant to ISO 7010.
Brand: Brady™ 829871
9.60 EUR valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Description
- Laminated polyester
- For indoor and outdoor use
Specifications
ISO 7010 | |
Gloss | |
3/Pk. | |
Indoor and Outdoor | |
0.099 mm | |
-40°C to 110°C | |
Global |
PIC W024-Tri 100-PE-SHEET/3 | |
Polyester | |
Permanent Acrylic | |
87 mm | |
100 mm | |
Triangular | |
Black on Yellow |