Learn More
Logistik™ Ladies Laboratory Coats
Includes three pockets. Logistik™ Ladies Laboratory Coats are used for a range of applications.
456.61 NOK
Specifications
Product Type | General Purpose Lab Coat |
---|---|
Gender | Women's |
Material | Polycotton |
Color | White |
Closure Type | Press Studs |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13192180
|
Logistik™
607-V3WH-100 |
16 | |||||||
Description
- With two lower pockets and breast pocket
- Stud fastened cuff
- Concealed stud fastened front
- Polycotton
- Not CE marked
Specifications
General Purpose Lab Coat | |
Polycotton | |
Press Studs | |
Reusable | |
3 |
Women's | |
White | |
Stud-fastened | |
Non-sterile | |
107 cm |
Product Title
Logistik™ Ladies Laboratory Coats