Learn More
Labconco™ Freeze-Drying Ampule Accessories
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
580.00 NOK - 15445.00 NOK
Specifications
For Use With (Equipment) Freeze drying system
View More Specs
Products 3
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11814750
View Documents
Labconco™
7576210
Stopper
580.00 NOK
Pack of 100
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 01-10-2025
Add to basket
15123075
View Documents
Labconco™
7578500
Sealing Torch
15445.00 NOK
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 12-01-2026
Add to basket
10248014
View Documents
Labconco™
7593401
Ampule Adapter
1670.00 NOK
Pack of 10
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 12-01-2026
Add to basket
Description
Description
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
  • Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
    • Package of 100.
  • Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
    • Seals all types of heat-resistant glass.
    • Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
  • Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
    • Ten per package.
Specifications
Specifications
Freeze drying system