Learn More
Labconco™ Atmospure Cable
Oxygen/Moisture monitor cable for use with Labconco Protector Glove Boxes
1990.00 NOK
Specifications
For Use With (Equipment) | AtmosPure |
---|---|
Weight (English) | 2 lb. |
Weight (Metric) | 0.9 Kg |
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Product Type | Price | Quantity & Availability | ||||
15977590
|
Labconco™
5442000 |
For use when Atmospure and glove box with auto pressure controller are sharing one vacuum pump | Atmospure 6-Pin Cable |
1990.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AtmosPure 9-Pin Oxygen/Moisture Monitor Cable is required for use when the AtmosPure is used in AUTO mode. Allows for automatic control of the AtmosPure blower from an accessory Oxygen or Moisture Monitor (sold separately).Specifications
AtmosPure | |
2 lb. | |
0.9 Kg |