Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
Brand:  DWK Life Sciences 774240-0014
261.35 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15327858
  • 342.00 NOK / Pack of 100
Only null left
Estimated Shipment: 29-10-2025
Description
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
  • Leak-proof, air-tight seals
  • Wide top flange for ease of use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14/10, 14/20, 14/35
Polyethylene
Plug Stopper