Learn More
Kinematica Polytron™ 2500E Homogenizer
Versatile units combine quality, convenience and high performance for rapid, reproducible sample processing and consistent sample-to-sample uniformity
Brand:  Kinematica 11010055
2089.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10785812
  • 2720.00€ / Each
Only null left
Estimated Shipment: 02-09-2025
Add to basket
Includes:
  • upport stand
Requires : Generators (sold separately; see Polytron Accessories)
Description
Description
  • Variable speed control
  • For heavy-duty use
  • High-speed, high-performance motor
  • Stand included
  • Soft-start motor prevents splashing and sample loss
  • Quick-connect: change generator with one hand
  • Accept wide range of stainless-steel Polytron generators (order separately)
Pulverizing plant and animal tissues, grains and seeds; soil extractions; DNA/RNA extractions, Karl Fischer sample prep, toxicology and food science
Specifications
Specifications
230 V
PT 2500 E