Learn More
DWK Life Sciences Kimble™ Kontes™ Parts for Microfiltration Assemblies
For use with ULTRA-WARE™ microfiltration assembly
4230.00 NOK - 9320.00 NOK
Specifications
For Use With (Application) | ULTRA-WARE microfiltration assembly |
---|---|
Quantity | Case of 1 |
Product Code | Brand | Material | Type | Diameter (Metric) Disc | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Type | Diameter (Metric) Disc | Volume (Metric) | Price | Quantity & Availability | ||
11873123
|
DWK Life Sciences
953810-0090 |
Stainless Steel | Support Screen | 90 mm |
9320.00 NOK
Each |
Estimated Shipment: 23-12-2025
Log in to see stock. |
|||
11708552
|
DWK Life Sciences
953781-90 |
Glass | Funnel | 90 mm | 1000 mL |
4230.00 NOK
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
||
Description
For use with ULTRA-WARE™ microfiltration assembly
Specifications
ULTRA-WARE microfiltration assembly | |
Case of 1 |