Learn More
3M™ Jupiter™ Ready-To-Use Starter Kits
Click to view available options
:
Model: RTUJUPITER; Filter type: A2P
Description
- High comfort decontamination belt
- Air Flow Indicator and Calibration tube
- Pair of Organic Vapour and Particulate (A2P) Filters and Prefilters
- Length-adjusting breathing tube
- 8-hour rechargeable battery (NiMH) and a battery charger
Specifications
Specifications
Battery Life | 8 hours |
Battery Type | NiMH |
Filter Type | OV/HE Filter |
Product Type | PAPR Assembly, No Headtop |
For Use With (Application) | Protection against gases, vapors, and particulates, or a combination of contaminants per filter used |
Includes | Powered turbo unit, decontamination belt, calibration tube and airflow indicator, 2 x A2P filters, pre-filters, breathing tube, rechargeable battery, battery charger |
Model | JPRTU1 |
Quantity | 1 Ea. |