Learn More
Cole-Parmer™ Jenway™ Glass Cuvettes
Brand:  Cole-Parmer™ 99610-32
 View more versions of this product
Product Code. 11480569
  • 3180.00 NOK / Each
Only null left
Estimated Shipment: 17-10-2025
Specifications
Cuvette
17.5 mL
Glass
50 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Jenway™ Glass Cuvettes > Capacity: Up to 17.5mL