Learn More
Jenway™ Buffer pH 10, Blue
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
Color | Blue |
For Use With (Equipment) | pH Electrode |
pH | 10 |
Quantity | 500 mL |
Packaging | Bottle |
Traceability to NIST | Yes |