Learn More
Cole-Parmer™ Jenway™ 100 Disposable Cuvettes
Brand: Cole-Parmer™ 99610-34
779.00 NOK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Cuvet | |
1.5 to 3.0 mL | |
100/Pk. | |
Spectral Measurement of Liquids |
Visible wavelengths only | |
Plastic | |
10 mm |