Learn More
Isopropyl-β-D-Thiogalactopyranoside, >99%, MP Biomedicals
Molecular Biology Reagent. Cell culture media component for use in molecular genetics.β-Galactoside inducer
518.00 NOK - 5775.00 NOK
Chemical Identifiers
CAS | 367-93-1 |
---|---|
Molecular Formula | C9H18O5S |
Molecular Weight (g/mol) | 238.30 |
MDL Number | MFCD00063273 |
InChI Key | BPHPUYQFMNQIOC-NXRLNHOXSA-N |
Synonym | iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside |
PubChem CID | 656894 |
ChEBI | CHEBI:61448 |
SMILES | CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11436511
|
MP Biomedicals
0219402980 |
100 mg |
518.00 NOK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11446511
|
MP Biomedicals
0219402983 |
250 mg |
544.00 NOK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460572
|
MP Biomedicals
0219402991 |
1 g |
1610.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470572
|
MP Biomedicals
0219402994 |
5 g |
5775.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480572
|
MP Biomedicals
0219402995 |
10 g |
5595.00 NOK
10g Qty Price Break Available |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
367-93-1 | |
238.30 | |
BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
656894 | |
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
C9H18O5S | |
MFCD00063273 | |
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
CHEBI:61448 |
Specifications
367-93-1 | |
C9H18O5S | |
iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
238.30 | |
CHEBI:61448 | |
Isopropyl-β-D-Thiogalactopyranoside |
>99% | |
MFCD00063273 | |
BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol | |
656894 | |
238.3 |
Safety and Handling
Recommended Storage : 0°C
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Isopropyl-beta-D-thiogalactopyranoside, molecular biology reagent
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl-lactopyranoside