Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
Brand: Mettler Toledo™ 30281926
906.66 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Detachable
- Resuable
Specifications
S7-None | |
To connect Mettler Toledo InLab sensors to any meter | |
3.0 m |
Mettler Toledo InLab Sensors | |
9.8 ft. |